2-(Aminooxy)-1-{4-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]piperazino}-1-ethanone hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:321391-87-1
:
Formula:C₁₂H₁₅Cl₂F₃N₄O₂
MDL Number:MFCD01114567
:375.18 g/mol
Melting Pt:172…177 °C
Storage Temperature:Ambient
Frequently Bought Together