
3-Fluoro-5-(4-morpholinylcarbonyl)phenylboronic acid 98%
Catalog # APOSPC4619-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:253.04 g/mol
Melting Pt:145…147 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:874219-40-6
Formula:C₁₁H₁₃BFNO₄
MDL Number:MFCD08235057