
5-Amino-1-(tert-butyl)-3-(4-fluorophenyl)-1H-pyrazole-4-carbonitrile 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:320422-34-2
:
Formula:C₁₄H₁₅FN₄
MDL Number:MFCD01568265
:258.3 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together