
2-Methoxy-4-(trifluoromethyl)benzonitrile 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:132927-08-3
:
Formula:C₉H₆F₃NO
MDL Number:MFCD02101784
Melting Pt:63…64 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together