
(2-Fluorophenyl)methanesulphonyl chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:24974-71-8
:
Formula:C₇H₆ClFO₂S
MDL Number:MFCD04117455
Melting Pt:54.5…56 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together