
2-Piperazin-1-yl-4-(trifluoromethyl)pyrimidine
Catalog # APOSPC4564-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.21 g/mol
Melting Pt:50…52 °C
Storage Temperature:Ambient
UN:3259
ADR:8,III
:GHS05
CAS Number:179756-91-3
Formula:C₉H₁₁F₃N₄
MDL Number:MFCD00203912