Order Entry
Czech Republic
ContactUsLinkComponent
(5-Fluoro-2-(trifluoromethyl)phenyl)acetic acid 98%
Irritant
(5-Fluoro-2-(trifluoromethyl)phenyl)acetic acid 98%
Catalog # APOSPC4557-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(5-Fluoro-2-(trifluoromethyl)phenyl)acetic acid 98%
Catalog # APOSPC4557-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :222.14 g/mol
Melting Pt:112…115 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:239135-52-5
Formula:C₉H₆F₄O₂
MDL Number:MFCD00236304