
6-(Trifluoromethyl)thionicotinamide 95+%
Catalog # APOSPC4545-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:206.19 g/mol
Melting Pt:160…161 °C
:GHS07
Storage Temperature:Ambient
CAS Number:386715-34-0
Formula:C₇H₅F₃N₂S
MDL Number:MFCD02173905