

2-Fluoro-6-(trifluoromethyl)phenol 98%
Catalog # APOSPC4539-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:180.1 g/mol
Melting Pt:35…36 °C
Storage Temperature:Refrigerator
UN:2924
ADR:3+8,III
:GHS05
:GHS07
CAS Number:239135-50-3
Formula:C₇H₄F₄O
MDL Number:MFCD00236300