1-[2-(4-Fluorophenoxy)ethyl]piperazine
Catalog # APOSPC449011-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:224.28 g/mol
Storage Temperature:Ambient
CAS Number:77602-92-7
Formula:C₁₂H₁₇FN₂O
MDL Number:MFCD04211513