

2-Bromo-1-(4-fluoro-3-methylphenyl)ethanone
Catalog # APOSPC446075-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:231.07 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
:GHS07
CAS Number:63529-31-7
Formula:C₉H₈BrFO
MDL Number:MFCD18089176