4-(6-Dimethylamino-4-trifluoromethylpyridin-2-yl)benzoic acid methyl ester
Catalog # APOSPC446063-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:324.31 g/mol
Storage Temperature:Ambient
CAS Number:1299607-60-5
Formula:C₁₆H₁₅F₃N₂O₂
MDL Number:MFCD18384852