Order Entry
Czech Republic
ContactUsLinkComponent
2-Bromo-5-fluoro-4-methylpyridine 98%
2-Bromo-5-fluoro-4-methylpyridine 98%
Catalog # APOSPC445054-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Bromo-5-fluoro-4-methylpyridine 98%
Catalog # APOSPC445054-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :190.02 g/mol
Storage Temperature:Ambient
CAS Number:885168-20-7
Formula:C₆H₅BrFN
MDL Number:MFCD08277310