Order Entry
Czech Republic
ContactUsLinkComponent
(S)-(+)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate
Irritant
(S)-(+)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate
Catalog # APOSPC440005-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(S)-(+)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate
Catalog # APOSPC440005-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :772.49 g/mol
Melting Pt:162…175 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:878111-17-2
Formula:C₃₆H₁₇F₁₂O₄P
MDL Number:MFCD08689863