


3-Fluoro-5-(trifluoromethyl)phenol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:172333-87-8
:
Formula:C₇H₄F₄O
MDL Number:MFCD00061288
:180.1 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
:GHS07
:GHS09
Frequently Bought Together