
2-Fluoro-6-(trifluoromethyl)benzyl bromide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:239087-08-2
:
Formula:C₈H₅BrF₄
MDL Number:MFCD00082477
:257.03 g/mol
Boiling Pt:45 °C/7.5mmHg
Melting Pt:38…45 °C
Flash Pt:107 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together