Order Entry
Czech Republic
ContactUsLinkComponent
2-Fluoro-5-(trifluoromethyl)benzylamine 97%
Corrosive
Irritant
2-Fluoro-5-(trifluoromethyl)benzylamine 97%
Catalog # APOSPC4374V-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
2-Fluoro-5-(trifluoromethyl)benzylamine 97%
Catalog # APOSPC4374V-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.14 g/mol
Storage Temperature:Ambient
UN:2735
ADR:8,III
 :GHS05
 :GHS07
CAS Number:199296-61-2
Formula:C₈H₇F₄N
MDL Number:MFCD00061162