
4-Fluoro-3-(trifluoromethyl)benzoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:67515-56-4
:
Formula:C₈H₃ClF₄O
MDL Number:MFCD00061159
Boiling Pt:169…171 °C
Density:1.493 g/cm³ (25 °C)
Flash Pt:76 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:226.56 g/mol
:GHS05
Frequently Bought Together