
3-Fluoro-5-(trifluoromethyl)benzoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:171243-30-4
:
Formula:C₈H₃ClF₄O
MDL Number:MFCD00061157
:226.56 g/mol
Boiling Pt:119 °C
Density:1.482 g/cm³ (25 °C)
Flash Pt:74 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together