
2-Fluoro-5-(trifluoromethyl)benzonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4088-84-0
:
Formula:C₈H₃F₄N
MDL Number:MFCD00061280
:189.11 g/mol
Boiling Pt:186 °C
Density:1.38 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
:GHS07
Frequently Bought Together