
2-Fluoro-4-(trifluoromethyl)benzonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:146070-34-0
:
Formula:C₈H₃F₄N
MDL Number:MFCD00061279
:189.11 g/mol
Boiling Pt:178…180 °C
Density:1.358 g/cm³ (25 °C)
Flash Pt:75 °C
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
:GHS07
Frequently Bought Together