
2-Fluoro-3-(trifluoromethyl)benzonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:146070-35-1
:
Formula:C₈H₃F₄N
MDL Number:MFCD00061152
Density:1.323 g/cm³ (25 °C)
Flash Pt:86 °C
UN:3276
ADR:6.1,III
:189.11 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together