
2-Fluoro-5-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:146137-78-2
:
Formula:C₈H₄F₄O
MDL Number:MFCD00061311
:192.11 g/mol
Boiling Pt:176 °C
Density:1.36 g/cm³ (25 °C)
Flash Pt:76 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together