
4'-Fluoro-2'-(trifluoromethyl)acetophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:208173-21-1
:
Formula:C₉H₆F₄O
MDL Number:MFCD00061251
:206.14 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together