
2'-Fluoro-6'-(trifluoromethyl)acetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:174013-29-7
:
Formula:C₉H₆F₄O
MDL Number:MFCD00061249
:206.14 g/mol
Density:1.326 g/cm³ (25 °C)
Flash Pt:86 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together