
2'-Fluoro-3'-(trifluoromethyl)acetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:207853-63-2
:
Formula:C₉H₆F₄O
MDL Number:MFCD00061145
:206.14 g/mol
Density:1.02 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together