

p,α,α,α-Tetrafluoroanisole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:352-67-0
:
Formula:C₇H₄F₄O
MDL Number:MFCD00040835
:180.1 g/mol
Boiling Pt:106 °C
Density:1.323 g/cm³ (25 °C)
Flash Pt:15 °C
Storage Temperature:Ambient
EINECS:206-523-2
UN:1993
ADR:3,II
:GHS02
:GHS07
Frequently Bought Together