
2,2-Difluoroethyltriethylammoniumtrifluoromethanesulphonate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:887267-07-4
:
Formula:C₉H₁₈F₅NO₃S
MDL Number:MFCD04038360
:315.3 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together