
Methyl-3-(trifluoromethoxy)benzoate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:148438-00-0
:
Formula:C₉H₇F₃O₃
MDL Number:MFCD00041005
:220.15 g/mol
Boiling Pt:197…199 °C
Density:1.445 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together