Order Entry
Czech Republic
ContactUsLinkComponent
4,6-Dichloro-2-(trifluoromethyl)pyrimidin-5-amine
4,6-Dichloro-2-(trifluoromethyl)pyrimidin-5-amine
Catalog # APOSPC430525-100MG
Supplier:  Apollo Scientific
CAS Number:  
4,6-Dichloro-2-(trifluoromethyl)pyrimidin-5-amine
Catalog # APOSPC430525-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :231.99 g/mol
Storage Temperature:Ambient
CAS Number:2344-17-4
Formula:C₅H₂Cl₂F₃N₃
MDL Number:MFCD08436613