2-[(4-Fluorobenzyl)amino]ethanol
Catalog # APOSPC430520-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:169.2 g/mol
Storage Temperature:Ambient
CAS Number:22116-33-2
Formula:C₉H₁₂FNO
MDL Number:MFCD04560903