
4'-Chloro-2'-fluoroacetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175711-83-8
:
Formula:C₈H₆ClFO
MDL Number:MFCD04116328
:172.59 g/mol
Flash Pt:95 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together