4,4-Difluorocyclohexanecarboxaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:265108-36-9
:
Formula:C₇H₁₀F₂O
MDL Number:MFCD09263468
:148.15 g/mol
Boiling Pt:55…57 °C/6mmHg
Storage Temperature:Freezer
UN:1989
ADR:3,III
Frequently Bought Together