

Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent) 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2356-16-3
:
Formula:C₈H₁₆FO₅P
MDL Number:MFCD00134400
:242.19 g/mol
Boiling Pt:75 °C/0.01mmHg
Density:1.194 g/cm³ (25 °C)
Flash Pt:73 °C
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS06
:GHS08
Frequently Bought Together