

1-Fluoro-4-(trifluoromethylthio)benzene 98%
Catalog # APOSPC4176-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:196.17 g/mol
Boiling Pt:136…138 °C
Flash Pt:37 °C
:GHS02
:GHS07
Storage Temperature:Refrigerator
CAS Number:940-76-1
Formula:C₇H₄F₄S
MDL Number:MFCD00040978