Order Entry
Czech Republic
ContactUsLinkComponent
(2-Fluorophenyl)acetonitrile 97%
Flammable
Irritant
(2-Fluorophenyl)acetonitrile 97%
Catalog # APOSPC4142-100G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
(2-Fluorophenyl)acetonitrile 97%
Catalog # APOSPC4142-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :135.14 g/mol
Boiling Pt:235 °C
Density:1.14 g/cm³ (25 °C)
Flash Pt:43 °C
Storage Temperature:Ambient
EINECS:206-313-0
UN:1992
ADR:3+6.1,III
 :GHS02
 :GHS07
CAS Number:326-62-5
Formula:C₈H₆FN
MDL Number:MFCD00001897