4-(Trifluoromethyl)phenylboronic acid pinacol ester 97%
Catalog # APOSPC412070-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:272.0712 g/mol
Storage Temperature:Ambient
CAS Number:214360-65-3
Formula:C₁₃H₁₆BF₃O₂
MDL Number:MFCD05863924