Order Entry
Czech Republic
ContactUsLinkComponent
 
(3-Chloro-4-ethoxy-2-fluorophenyl)boronic acid 98%
Catalog # APOSPC412029-5G
Supplier:  Apollo Scientific
CAS Number:  
(3-Chloro-4-ethoxy-2-fluorophenyl)boronic acid 98%
Catalog # APOSPC412029-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :218.4177 g/mol
Storage Temperature:Ambient
CAS Number:909122-50-5
Formula:C₈H₉BClFO₃
MDL Number:MFCD15144702