Order Entry
Czech Republic
ContactUsLinkComponent
3-(4-Fluorophenoxy)propionic acid 97%
Irritant
3-(4-Fluorophenoxy)propionic acid 97%
Catalog # APOSPC4113-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-(4-Fluorophenoxy)propionic acid 97%
Catalog # APOSPC4113-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:83…85 °C
EINECS:216-424-6
 :GHS07
Storage Temperature:Ambient
CAS Number:1579-78-8
Formula:C₉H₉FO₃
MDL Number:MFCD00052838