
2-(3-Trifluoromethylpyrazol-1-yl)ethylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1006436-51-6
:
Formula:C₆H₈F₃N₃
MDL Number:MFCD04970067
:179.15 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together