Order Entry
Czech Republic
ContactUsLinkComponent
 
1-(Difluoromethyl)-3-methyl-1H-pyrazole-5-carboxylic acid
Catalog # APOSPC410370-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(Difluoromethyl)-3-methyl-1H-pyrazole-5-carboxylic acid
Catalog # APOSPC410370-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :176.12 g/mol
Storage Temperature:Ambient
CAS Number:1006319-91-0
Formula:C₆H₆F₂N₂O₂
MDL Number:MFCD04970129