4-[4-(Trifluoromethyl)phenyl]piperidine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:574008-73-4
:
Formula:C₁₂H₁₅ClF₃N
MDL Number:MFCD02178912
:265.71 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together