tert-Butyl-4-(2-(trifluoromethyl)-1,6-naphthyridin-5-ylamino)piperidine-1-carboxylate 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1040682-38-9
:
Formula:C₁₉H₂₃F₃N₄O₂
MDL Number:MFCD10688282
:396.42 g/mol
Storage Temperature:Ambient
Frequently Bought Together