Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 7-Chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate 95+%
Ethyl 7-Chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate 95+%
Catalog # APOSPC408427-25G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 7-Chloro-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate 95+%
Catalog # APOSPC408427-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :382.72 g/mol
Storage Temperature:Ambient
CAS Number:100491-29-0
Formula:C₁₇H₁₀ClF₃N₂O₃
MDL Number:MFCD01863285