3-Fluoro-5-(pentafluorothio)benzylamine 98%
Catalog # APOSPC405665-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:251.19 g/mol
Storage Temperature:Ambient
CAS Number:1240257-73-1
Formula:C₇H₇F₆NS
MDL Number:MFCD16652463