Order Entry
Czech Republic
ContactUsLinkComponent
3-Bromo-5-(pentafluorothio)benzyl bromide 97%
Corrosive
3-Bromo-5-(pentafluorothio)benzyl bromide 97%
Catalog # APOSPC405658-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
3-Bromo-5-(pentafluorothio)benzyl bromide 97%
Catalog # APOSPC405658-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :375.98 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
 :GHS05
CAS Number:1240257-14-0
Formula:C₇H₅Br₂F₅S
MDL Number:MFCD16652484