(DL)-4-(trans-2-(Trifluoromethyl)cyclopropyl)benzoic acid
Catalog # APOSPC405603-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:177.2 g/mol
Storage Temperature:Ambient
CAS Number:1404192-14-8
Formula:C₁₀H₁₁NO₂