
(3,3-Difluorocyclobutyl)methanol
Catalog # APOSPC405001-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:122.12 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:681128-39-2
Formula:C₅H₈F₂O
MDL Number:MFCD11877861