Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-3-(chloromethyl)-7-fluoroquinoline
2-Chloro-3-(chloromethyl)-7-fluoroquinoline
2-Chloro-3-(chloromethyl)-7-fluoroquinoline
APOSPC404555-1G
:
Apollo Scientific
:
2-Chloro-3-(chloromethyl)-7-fluoroquinoline
APOSPC404555-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
Ambient
:
948291-38-1
:
C₁₀H₆Cl₂FN