2-Chloro-3-(chloromethyl)-6-fluoroquinoline
Catalog # APOSPC404554-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:948291-04-1
Formula:C₁₀H₆Cl₂FN